Skip to Content
Merck
All Photos(1)

Key Documents

V800297

Sigma-Aldrich

Petroleum ether

AR, bp 60-80 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
grade:
AR
bp:
30-60 °C (lit.)
60-80 °C
vapor pressure:
25.8 psi ( 55 °C)
7.99 psi ( 20 °C)
Product V800297 is not currently sold in your country. Contact Technical Service

grade

AR

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

product line

Vetec

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

dilution

(for analytical testing)

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)
60-80 °C

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Petroleum ether, a mixture of aliphatic hydrocarbons, is majorly used as a laboratory solvent. It mainly contains pentane and hexane.[1][2] Petroleum ether has properties similar to ether and is used in cosmetics, detergents, paint, and varnish, etc.[3][4]

Application

Petroleum ether can be used:     
  • As a solvent in API production.[5]       
  • As a chromatography solvent for product extraction, and purification of non-polar compounds.[6]       
  • In the extraction of oil, wax and fat.[7]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of moisture content and temperature on mechanical extraction of oil from watermelon (Citrullus lanatus) seeds
Poornima DS, et al.
Journal of Pharmacognosy and Phytochemistry, 8(4), 275-279 (2019)
Solvent efficiency for oil extraction from spent bleaching clay
Lee CG, et al.
Journal of the American Oil Chemists' Society, 77(11), 1219-1223 (2000)
Isolation of stigmasterol and β -sitosterol from petroleum ether extract of aerial parts of Ageratum conyzoides (Asteraceae)
Kamboj A and Saluja AK
International Journal of Pharmacy and Pharmaceutical Sciences, 3(1), 94-96 (2011)
XLII.?Some hydrocarbons from american petroleum. I. Normal and iso-pentane
Young S and Thomas GL
Journal of the Chemical Society, 71, 440-446 (1897)
Antibacterial activities of bee venom, propolis, and royal jelly produced by three honey bee, Apis mellifera L., hybrids reared in the same environmental conditions
Attalla KM, et al.
Annals of Agricultural Science, 45, 895-902 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service