Skip to Content
Merck
All Photos(1)

Key Documents

V800295

Sigma-Aldrich

Petroleum ether

AR, bp 40-60 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
grade:
AR
bp:
30-60 °C (lit.)
40-60 °C
vapor pressure:
25.8 psi ( 55 °C)
7.99 psi ( 20 °C)
Product V800295 is not currently sold in your country. Contact Technical Service

grade

AR

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

product line

Vetec

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)
40-60 °C

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Petroleum ether is generally a mixture of low boiling aliphatic hydrocarbons mainly hexane and pentane.[1] It is generally used as a laboratory solvent[1] and also as an extraction solvent.[2]

Application

Iron(II) acetate may be used as an iron precursor for the synthesis of phase-pure iron carbide nanoparticles with potential applications in the field of biomedicine and catalysis due to its high magnetic saturation and chemical stability.[3]

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F - closed cup

Flash Point(C)

-49 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of monocarboxylic iron derivatives in the ring-opening polymerization of L-lactide.
Stolt M & Soderg?rd A.
Macromolecules, 32(20), 6412-6417 (1999)
Microwave-assisted extraction of artemisinin from Artemisia annua L.
Hao J-Y, et al.
Separation and Purification Technology, 28(3), 191-196 (2002)
CRC Handbook of Chemistry and Physics, 85 (2004)
Lena Ernstgård et al.
Journal of applied toxicology : JAT, 29(3), 255-262 (2008-12-17)
A move from 'standard' white spirit (stdWS, 15-20% aromatics) to low-aromatic or dearomatized white spirit (deWS) has been seen, as the latter are considered to carry a smaller risk of health effects. However, data on health risks of deWS on
Gunnar Damgård Nielsen et al.
Basic & clinical pharmacology & toxicology, 98(2), 115-123 (2006-02-01)
Numerous studies have suggested that long-term occupational exposure to white spirit may cause chronic toxic encephalopathy (WHO 1996). This review summarizes the chronic nervous system effects of white spirit in animal studies during a 30-year period. First, routine histopathology was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service