Skip to Content
Merck
All Photos(1)

Key Documents

V800262

Sigma-Aldrich

Methanol

≥99.5%, anhydrous, Vetec

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
EC Number:
MDL number:
UNSPSC Code:
15101803
PubChem Substance ID:
grade:
anhydrous
Assay:
≥99.5%
bp:
64.7 °C (lit.)
vapor pressure:
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Product Name

Methanol, anhydrous, ≥99.5%

grade

anhydrous

vapor density

1.11 (vs air)

vapor pressure

410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

product line

Vetec

Assay

≥99.5%

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanol (carbinol or methyl alcohol or wood alcohol) is a primary aliphatic alcohol miscible in water. It′s a polar solvent used in many organic synthesis reactions. Recently, it has been used in direct methanol fuel cell (DMFC) technology and in the production of gasoline, olefins, and hydrocarbons.

Application

Methanol can be used in the:
  • Preparation of methyl N-phenyl carbamate using urea and aniline.
  • Preparation of biodiesel from rapeseed oil using metal oxide (catalyst).
  • Silanization of multi-walled carbon nanotubes.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One pot synthesis of methyl N-phenyl carbamate from aniline, urea and methanol
Qin F, et al.
Catalysis Letters, 126(3-4), 419-425 (2008)
Synthesis of biodiesel from rapeseed oil using supercritical methanol with metal oxide catalysts
Yoo S, et al.
Bioresource Technology, 101(22), 8686-8689 (2010)
Methanol
Young J A
Journal of Chemical Education, 83(8), 1131-1131 (2006)
Functionalization of carbon nanotubes using a silane coupling agent
Ma P, et al.
Carbon, 44(15), 3232-3238 (2006)
Direct conversion technologies of methane to methanol: an overview
Zakaria Z and Kamarudin S K
Renewable and Sustainable Energy Reviews, 65(8), 250-261 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service