Skip to Content
Merck
All Photos(1)

Key Documents

V800178

Sigma-Aldrich

2-Butanone

LR, ≥99%

Synonym(s):

Ethyl methyl ketone, MEK, Methyl ethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5COCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
741880
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
Assay:
≥99%
bp:
80 °C (lit.)
vapor pressure:
71 mmHg ( 20 °C)

grade

LR

vapor density

2.49 (vs air)

vapor pressure

71 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

960 °F

expl. lim.

10.1 %

refractive index

n20/D 1.379 (lit.)

bp

80 °C (lit.)

mp

−87 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CC(CC)=O

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L B Goldsmith et al.
Contact dermatitis, 19(5), 348-350 (1988-11-01)
Irritant contact dermatitis along with an increased transepidermal water loss can result from exposing the skin to solvents. A study of the interaction of various solvents with human stratum corneum was made using thin-layer chromatography. Comparison of 10 solvents (trichloroethylene
S G Carmella et al.
Cancer research, 53(4), 721-724 (1993-02-15)
Metabolites of the tobacco-specific nitrosamine, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone, a potent pulmonary carcinogen, have been quantified in the urine of 11 smokers. They were not detected in nonsmokers' urine. The metabolites, 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol and its glucuronide, were detected in quantities of 0.23-1.0 and 0.57-6.5
Jeong-Hwan Yoon et al.
Nature communications, 6, 7600-7600 (2015-07-22)
Transforming growth factor-β (TGF-β) and interleukin-6 (IL-6) are the pivotal cytokines to induce IL-17-producing CD4(+) T helper cells (TH17); yet their signalling network remains largely unknown. Here we show that the highly homologous TGF-β receptor-regulated Smads (R-Smads): Smad2 and Smad3
Sherille D Bradley et al.
Cancer immunology research, 3(6), 602-609 (2015-03-22)
Oncogene activation in tumor cells induces broad and complex cellular changes that contribute significantly to disease initiation and progression. In melanoma, oncogenic BRAF(V600E) has been shown to drive the transcription of a specific gene signature that can promote multiple mechanisms
L Schafranek et al.
Leukemia, 29(1), 76-85 (2014-05-13)
Kinase inhibitors block proliferative signals in BCR-ABL1+ leukemic cells, but their capacity to induce apoptosis is poorly understood. Initial studies suggested that very brief exposure to kinase inhibitors was sufficient to induce apoptosis in chronic myeloid leukemia (CML) cells. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service