Skip to Content
Merck
All Photos(1)

Documents

V800133

Sigma-Aldrich

Copper(II) sulfate pentahydrate

LR, ≥99%

Synonym(s):

Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

vapor pressure

7.3 mmHg ( 25 °C)

product line

Vetec

Assay

≥99%

form

solid

pH

3.5-4.5 (20 °C, 50 g/L)

mp

110 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤0.05%

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azizallah Feizollahi et al.
Analytical methods : advancing methods and applications, 13(7), 910-917 (2021-01-30)
Determination and sensing of antibiotics in dairy products are the biggest challenges in the world. In continuation of our earlier study, a facile and novel determination method for the detection of sulfamethazine (SMZ) in cow milk has been developed using
Nikolaos Kontoudakis et al.
Food chemistry, 316, 126352-126352 (2020-02-18)
This study investigates the relationship between the two main forms of Cu in wine and their impact on the rate of oxygen consumption. The Cu forms were differentiated by medium exchange constant current stripping potentiometry, which classified the Cu as
Viktoriia Korniienko et al.
Materials (Basel, Switzerland), 13(18) (2020-09-10)
High strength, excellent corrosion resistance, high biocompatibility, osseointegration ability, and low bacteria adhesion are critical properties of metal implants. Additionally, the implant surface plays a critical role as the cell and bacteria host, and the development of a simultaneously antibacterial
Nikolaos Kontoudakis et al.
Food research international (Ottawa, Ont.), 98, 95-102 (2017-06-15)
A variety of techniques have been developed with the ability to measure different forms of metals in wine with the ultimate aim of providing a more accurate indicator of metal induced spoilage of wine. This study was conducted in order
Bhagwan S Batule et al.
International journal of nanomedicine, 10 Spec Iss, 137-142 (2015-09-09)
We developed a simple but efficient method to synthesize protein-inorganic hybrid nanostructures with a flower-like shape (nanoflowers), which relies on sonication to facilitate the synthesis of the nanoflowers. With this technique, we synthesized nanoflowers containing laccase as a model protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service