Skip to Content
Merck
All Photos(1)

Key Documents

V800070

Sigma-Aldrich

Benzaldehyde

AR, ≥99%

Synonym(s):

Bitter almond

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CHO
CAS Number:
Molecular Weight:
106.12
Beilstein:
471223
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

AR

Quality Level

vapor density

3.7 (vs air)

vapor pressure

4 mmHg ( 45 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

374 °F

expl. lim.

1.4 %, 20 °F

refractive index

n20/D 1.545 (lit.)

pH

5.9 (20 °C)

bp

178-179 °C (lit.)

mp

−26 °C (lit.)

density

1.044 g/cm3 at 20 °C (lit.)

SMILES string

O=Cc1ccccc1

InChI

1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H

InChI key

HUMNYLRZRPPJDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-gen Peng et al.
Chemical communications (Cambridge, England), 49(26), 2709-2711 (2013-02-26)
A center radially fibrous silica encapsulated TS-1 zeolite (TS-1@KCC-1) has been synthesized in a microemulsion system for the first time. Supporting the Rh(OH)3 species, this novel core-shell structured material serves as a robust bifunctional catalyst for one-pot synthesis of benzamide
Guanying Li et al.
Biomaterials, 63, 128-136 (2015-06-24)
Sulfur dioxide (SO2) and its derivatives sulfite and bisulfite play important roles in biological systems. However, in vivo detection of sulfite/bisulfite remains challenging. In this study, we developed a dinuclear Ir(III) complex (Ir4) as a two-photon phosphorescent probe for sulfite and
Longyun Lv et al.
ACS combinatorial science, 15(4), 183-192 (2013-02-22)
We previously reported the novel efficient proton/heat-promoted four-component reactions (4CRs) of but-2-ynedioates, two same/different primary amines, and aldehydes for the synthesis of tetra- and pentasubstituted polyfunctional dihydropyrroles. If aromatic and aliphatic amines were used as reagents, four different series of
Lidia Mateo et al.
PLoS genetics, 10(8), e1004560-e1004560 (2014-08-15)
The increase in availability of whole genome sequences makes it possible to search for evidence of adaptation at an unprecedented scale. Despite recent progress, our understanding of the adaptive process is still very limited due to the difficulties in linking
Mirjam Appel et al.
PloS one, 10(5), e0126986-e0126986 (2015-05-21)
Electric shock is a common stimulus for nociception-research and the most widely used reinforcement in aversive associative learning experiments. Yet, nothing is known about the mechanisms it recruits at the periphery. To help fill this gap, we undertook a genome-wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service