Skip to Content
Merck
All Photos(2)

Key Documents

V800061

Sigma-Aldrich

Antimony(III) chloride

LR, ≥98.5%

Synonym(s):

Antimony trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SbCl3
CAS Number:
Molecular Weight:
228.12
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystals
flakes
Product V800061 is not currently sold in your country. Contact Technical Service

grade

LR

vapor density

7.9 (vs air)

vapor pressure

1 mmHg ( 49 °C)

product line

Vetec

Assay

≥98.5%

form

crystals
flakes

mp

73.4 °C (lit.)

cation traces

Fe: ≤0.002%

SMILES string

Cl[Sb](Cl)Cl

InChI

1S/3ClH.Sb/h3*1H;/q;;;+3/p-3

InChI key

FAPDDOBMIUGHIN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Gurnani et al.
Cytobios, 70(281), 131-136 (1992-01-01)
Clastogenic effects of antimony trichloride, used in small industries, were monitored in laboratory bred white Swiss mice in vivo, following oral administration by gavaging, after 6, 12, 18 and 24 h. Chromosomal aberrations were principally breaks and damaged cells observed
T Gebel et al.
Anticancer research, 18(6A), 4253-4257 (1999-01-19)
Antimony compounds are supposed to resemble to arsenicals in some toxicological features. Comparative investigations with antimony and arsenic were performed to collect data on the genotoxicity of antimony, on which the knowledge is scarce. In comparison to trivalent arsenic, trivalent
The response of different photometers to the color produced by vitamin A and carotene with antimony trichloride.
M J CALDWELL et al.
Transactions of the Kansas Academy of Science. Kansas Academy of Science, 49(2), 197-204 (1946-09-01)
Why not Nichols chemosurgery?
Gary A Dyer
Clinics in dermatology, 24(5), 458-460 (2006-09-13)
T Gebel
Mutation research, 412(3), 213-218 (1998-05-26)
Arsenic and antimony are two semimetals sharing some chemical as well as toxicological properties. Both elements are clastogenic but not point mutagenic in their trivalent state of valency. Environmental exposure to arsenic was proven to be associated with increased rates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service