Skip to Content
Merck
All Photos(2)

Key Documents

V800045

Sigma-Aldrich

Ammonium iron(III) sulfate dodecahydrate

AR, ≥99%

Synonym(s):

Ammonium ferric sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Fe(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
482.19
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
crystalline powder

grade

AR

product line

Vetec

Assay

≥99%

form

crystalline powder

mp

39-41 °C (lit.)

anion traces

chloride (Cl-): ≤0.0005%

cation traces

Fe: ≤0.001%
K: ≤0.01%
Na: ≤0.01%
heavy metals (as Pb): ≤0.002%

SMILES string

N.O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

LCPUDZUWZDSKMX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Wittmann et al.
Chemico-biological interactions, 138(2), 171-187 (2001-10-24)
Diabetes mellitus is characterized by increased methylglyoxal (MG) production. The aim of the present study was to investigate the role of iron in the cellular and molecular effects of MG. A red blood cell (RBC) model and L-arginine were used
J G Reinhold et al.
The Journal of nutrition, 116(6), 1007-1017 (1986-06-01)
The effects of pH, ascorbate, citrate and dietary fiber on retention of ferrous and ferric iron by jejuno-ileal segments of rat intestine were examined in vivo. Iron was introduced in an isosmotic solution of sodium chloride and dextrose buffered by
Andrew J Ghio et al.
American journal of physiology. Lung cellular and molecular physiology, 283(3), L563-L572 (2002-08-10)
The iron chelator deferoxamine has been reported to inhibit both xanthine oxidase (XO) and xanthine dehydrogenase activity, but the relationship of this effect to the availability of iron in the cellular and tissue environment remains unexplored. XO and total xanthine
H S Kwak et al.
Journal of agricultural and food chemistry, 51(26), 7770-7774 (2003-12-11)
This study was designed to develop a microencapsulated iron that could be used to fortify milk and to determine the sensory properties of milk fortified with microencapsulated iron. Coating material was polyglycerol monostearate (PGMS), and selected core material was ferric
Varuni S Munasinghe et al.
Parasitology, 139(7), 864-869 (2012-02-18)
SUMMARYDientamoeba fragilis is an intestinal protozoan in humans that is commonly associated with diarrhoea and other gastrointestinal complaints. Studies conducted to investigate the biology of this parasite are limited by methods for in vitro cultivation. The objective of this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service