Skip to Content
Merck
All Photos(1)

Key Documents

V800023

Sigma-Aldrich

Acetone

AR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
AR
Assay:
≥99.5%
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)
Product V800023 is not currently sold in your country. Contact Technical Service

grade

AR

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

product line

Vetec

Assay

≥99.5%

form

liquid

expl. lim.

13.2 %

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetone may be used as a solvent in the synthesis of the bismuth oxide electrode, which shows reversible and stable redox reactions in different types of aqueous electrolytes, high rate capability, and good cycle life with potential applicability in aqueous rechargeable batteries.[1]
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bismuth oxide: a versatile high-capacity electrode material for rechargeable aqueous metal-ion batteries.
Zuo W, et al.
Energy & Environmental Science, 9(9), 2881-2891 (2016)
Hugo Lövheim et al.
Alzheimer's & dementia : the journal of the Alzheimer's Association, 11(6), 593-599 (2014-07-22)
Previous studies have suggested a link between herpes simplex virus (HSV) type 1 and the development of Alzheimer's disease (AD). The present analysis included 3432 persons (53.9% women, mean age at inclusion 62.7 ± 14.4 years) with a mean follow-up
Oleksiy Krupin et al.
Optics express, 21(1), 698-709 (2013-02-08)
Straight long-range surface plasmon waveguides are demonstrated as biosensors for the detection of cells, proteins and changes in the bulk refractive index of solutions. The sensors consist of 5 μm wide 22 nm thick Au stripes embedded in polymer (CYTOP™)
Juan A López-Ráez et al.
Plant science : an international journal of experimental plant biology, 230, 59-69 (2014-12-07)
Apocarotenoids are a class of compounds that play important roles in nature. In recent years, a prominent role for these compounds in arbuscular mycorrhizal (AM) symbiosis has been shown. They are derived from carotenoids by the action of the carotenoid
Wanqiang Sheng et al.
Cell research, 24(12), 1387-1402 (2014-11-22)
T helper (TH)-cell subsets, such as TH1 and TH17, mediate inflammation in both peripheral tissues and central nervous system. Here we show that STAT5 is required for T helper-cell pathogenicity in autoimmune neuroinflammation but not in experimental colitis. Although STAT5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service