Skip to Content
Merck
All Photos(1)

Key Documents

V001604

Sigma-Aldrich

Creatinine

98.5-102.0%

Synonym(s):

2-Amino-1-methyl-2-imidazolin-4-one, 2-Imino-1-methylimidazolidin-4-one, 2-Imino-N-methylhydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N3O
CAS Number:
Molecular Weight:
113.12
Beilstein:
112061
EC Number:
MDL number:
PubChem Substance ID:

Quality Level

Assay

98.5-102.0%

mp

295 °C (dec.) (lit.)

SMILES string

CN1CC(=O)N=C1N

InChI

1S/C4H7N3O/c1-7-2-3(8)6-4(7)5/h2H2,1H3,(H2,5,6,8)

InChI key

DDRJAANPRJIHGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

554.0 °F - closed cup

Flash Point(C)

290.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven Orlov et al.
Clinical journal of the American Society of Nephrology : CJASN, 10(7), 1136-1144 (2015-06-21)
Cardiac autonomic neuropathy predicts future adverse renal outcomes in the general population. This study sought to determine its relationship with early progressive renal decline in type 1 diabetes. A subset of participants with normoalbuminuria (n=204) or microalbuminuria (n=166) from the
Amit X Garg et al.
JAMA, 311(21), 2191-2198 (2014-06-03)
Most acute kidney injury observed in the hospital is defined by sudden mild or moderate increases in the serum creatinine concentration, which may persist for several days. Such acute kidney injury is associated with lower long-term kidney function. However, it
Zoe K McQuilten et al.
The Journal of thoracic and cardiovascular surgery, 150(1), 216-222 (2015-05-06)
To assess whether introduction of universal leukodepletion (ULD) of red blood cells (RBCs) for transfusion was associated with improvements in cardiac surgery patient outcomes. Retrospective study (2005-2010) conducted at 6 institutions. Associations between leukodepletion and outcomes of mortality, infection, and
Sarah H Koning et al.
Kidney international, 87(5), 1009-1016 (2015-01-15)
There are few reports of associations between alcohol consumption and risk of chronic kidney disease (CKD). To investigate this further, we studied 5476 participants aged 28-75 years in the Prevention of Renal and Vascular End-Stage Disease (PREVEND) study, a prospective
Karel G M Moons et al.
Annals of internal medicine, 162(1), W1-73 (2015-01-07)
The TRIPOD (Transparent Reporting of a multivariable prediction model for Individual Prognosis Or Diagnosis) Statement includes a 22-item checklist, which aims to improve the reporting of studies developing, validating, or updating a prediction model, whether for diagnostic or prognostic purposes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service