Skip to Content
Merck
All Photos(1)

Key Documents

V000188

Sigma-Aldrich

Petroleum ether

bp 30-60 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12190000

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

autoignition temp.

475 °F

expl. lim.

8 %

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)
30-60 °C

density

0.64 g/mL at 25 °C (lit.)

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F

Flash Point(C)

-49 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Bondy et al.
Archives of toxicology, 69(6), 410-414 (1995-01-01)
Levels of glutathione and activity of glutamine synthetase were assayed in organs of rats following inhalation of a heterogeneous solvent mixture containing both aliphatic and aromatic hydrocarbons. This mixture was administered for 3 weeks (6 h daily) at two levels
I P Dick et al.
Human & experimental toxicology, 16(11), 645-651 (1998-01-14)
1. The absorption and distribution of lindane through skin was examined using human volunteers. Two different preparations were employed, one with acetone as the vehicle and the other, a commercial product, consisting primarily of white spirit as the solvent base.
The use of a surgical glove in the prevention of tourniquet-associated chemical burns.
P J Tomlinson et al.
Annals of the Royal College of Surgeons of England, 90(3), 255-255 (2008-04-26)
Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further
I P Dick et al.
Human & experimental toxicology, 16(11), 652-657 (1998-01-14)
1. The absorption of lindane through human skin was assessed in vitro using static diffusion cells with 50% aqueous ethanol as the receptor fluid. Four formulations of lindane were applied, three of which were commercial preparations. The dermal distribution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service