1A01650
USP
3-Hydroxydesloratadine
Pharmaceutical Analytical Impurity (PAI)
Synonym(s):
3-Hydroxy Desloratadine
Select a Size
About This Item
grade
pharmaceutical analytical impurity (PAI)
Agency
USP
API family
desloratadine
manufacturer/tradename
USP
application(s)
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
Clc1cc2c(cc1)C(=C4CCNCC4)c3ncc(cc3CC2)O
InChI
1S/C19H19ClN2O/c20-15-3-4-17-13(9-15)1-2-14-10-16(23)11-22-19(14)18(17)12-5-7-21-8-6-12/h3-4,9-11,21,23H,1-2,5-8H2
InChI key
NDFMTPISBHBIKE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.
Associated Drug Substance: Desloratadine
Therapeutic Area: Antihistamines.
For more information about this PAI, visit here.
Application
Features and Benefits
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service