Skip to Content
Merck
All Photos(1)

Key Documents

1681622

USP

Triclabendazole Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Triclabendazole sulfoxide, 5-Chloro-6-(2,3-dichlorophenoxy)-2-(methylsulfinyl)-1H-benzimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$2,220.00

$2,220.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
15 MG
$2,220.00

About This Item

Empirical Formula (Hill Notation):
C14H9Cl3N2O2S
CAS Number:
Molecular Weight:
375.66
Beilstein:
5829656
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$2,220.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

Agency

USP

API family

triclabendazole

packaging

pkg of 15 mg

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

CS(=O)c1nc2cc(Cl)c(Oc3cccc(Cl)c3Cl)cc2[nH]1

InChI

1S/C14H9Cl3N2O2S/c1-22(20)14-18-9-5-8(16)12(6-10(9)19-14)21-11-4-2-3-7(15)13(11)17/h2-6H,1H3,(H,18,19)

InChI key

GABQPFWIQFRJSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Triclabendazole Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Triclabendazole

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service