1597504
USP
Quininone
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(8α)-6′-Methoxycinchonan-9-one
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
quinine
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
InChI
1S/C20H22N2O2/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18/h3-6,8,11,13-14,19H,1,7,9-10,12H2,2H3
InChI key
SRFCUPVBYYAMIL-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Quinidine Sulfate
- Quinine Sulfate
- Quinidine Gluconate
- Quinidine Gluconate Injection
- Quinidine Sulfate Tablets
- Quinine Sulfate Capsules
- Quinine Sulfate Tablets
Analysis Note
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service