1571001
USP
Proparacaine hydrochloride
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Proxymetacaine hydrochloride
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
proparacaine
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Cl.CCCOc1ccc(cc1N)C(=O)OCCN(CC)CC
InChI
1S/C16H26N2O3.ClH/c1-4-10-20-15-8-7-13(12-14(15)17)16(19)21-11-9-18(5-2)6-3;/h7-8,12H,4-6,9-11,17H2,1-3H3;1H
InChI key
BFUUJUGQJUTPAF-UHFFFAOYSA-N
Gene Information
human ... SCN10A(6336) , SCN11A(11280) , SCN1A(6323) , SCN2A(6326) , SCN3A(6328) , SCN4A(6329) , SCN5A(6331) , SCN7A(6332) , SCN8A(6334) , SCN9A(6335)
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Proparacaine Hydrochloride Ophthalmic Solution
- Fluorescein Sodium and Proparacaine Hydrochloride Ophthalmic Solution
Analysis Note
Other Notes
related product
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service