Skip to Content
Merck
All Photos(1)

Key Documents

1554148

USP

Prasugrel Related Compound D

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-5,6,7,7a-tetrahydrothieno[3,2-c]pyridin-2(4H)-one)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18FNO2S
CAS Number:
Molecular Weight:
331.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

USP

API family

prasugrel

manufacturer/tradename

USP 1554148

application(s)

pharmaceutical

format

neat

storage temp.

room temp

SMILES string

Fc1c(cccc1)C(N3CCC4SC(=O)C=C4C3)C(=O)C2CC2

InChI

1S/C18H18FNO2S/c19-14-4-2-1-3-13(14)17(18(22)11-5-6-11)20-8-7-15-12(10-20)9-16(21)23-15/h1-4,9,11,15,17H,5-8,10H2

InChI key

MJAMUSZUMAHFLH-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis. USP issued SDS can be found here.

Application

Prasugrel Related Compound D USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monograph such as Prasugrel Hydrochloride.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service