Skip to Content
Merck
All Photos(1)

Key Documents

1544417

USP

Piroctone Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-[5-Methyl-3-(2,4,4-trimethylpentyl)-4,5-dihydroisoxazol-5-yl]acetic acid 2-aminoethanol salt

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
$961.00

$961.00


Estimated to ship on23 March 2025


Request a Bulk Order

Select a Size

Change View
15 MG
$961.00

About This Item

Empirical Formula (Hill Notation):
C16H32N2O4
Molecular Weight:
316.44
UNSPSC Code:
41116107
NACRES:
NA.24

$961.00


Estimated to ship on23 March 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

piroctone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Piroctone Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service