Skip to Content
Merck
All Photos(1)

Key Documents

1529016

USP

Phentolamine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-(2-Aminoethyl)-2-[(3-hydroxyphenyl)(4-tolyl)amino]acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$2,290.00

$2,290.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$2,290.00

About This Item

Empirical Formula (Hill Notation):
C17H21N3O2
Molecular Weight:
299.37
UNSPSC Code:
41116107
NACRES:
NA.24

$2,290.00


Check Cart for Availability

Request a Bulk Order

API family

phentolamine

packaging

pkg of 25 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Phentolamine Related Compound A USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Phentolamine Mesylate
  • Phentolamine Mesylate Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service