Skip to Content
Merck
All Photos(1)

Key Documents

1526007

USP

Phenoxybenzamine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-(2-Chloroethyl)-N-(1-methyl-2-phenoxyethyl)benzylamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$696.00

$696.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$696.00

About This Item

Empirical Formula (Hill Notation):
C18H22ClNO · HCl
CAS Number:
Molecular Weight:
340.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$696.00


Estimated to ship on16 April 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

phenoxybenzamine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

15-25°C

SMILES string

Cl[H].CC(COc1ccccc1)N(CCCl)Cc2ccccc2

InChI

1S/C18H22ClNO.ClH/c1-16(15-21-18-10-6-3-7-11-18)20(13-12-19)14-17-8-4-2-5-9-17;/h2-11,16H,12-15H2,1H3;1H

InChI key

VBCPVIWPDJVHAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Calmodulin antagonist; α-adrenoceptor antagonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Telegdy et al.
Behavioural brain research, 281, 333-338 (2014-12-30)
The action of PACAP-38 was studied by measuring the anxiogenic-anxiolytic behavior of rats in an elevated plus maze. PACAP-38 was administered into the lateral brain ventricle and the behavior of the animals was measured 3h later. The possible involvement of
G Telegdy et al.
Behavioural brain research, 270, 326-329 (2014-05-13)
Urocortin 3 (Ucn 3) was found to impair passive avoidance learning in male and female mice. The possible involvement of transmitters in the action, the animals were pretreated with the following receptor antagonists in doses which alone could not influence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service