Skip to Content
Merck
All Photos(1)

Key Documents

1451000

USP

Nalidixic acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,4-Dihydro-1-ethyl-7-methyl-1,8-naphthyridin-4-one-3-carboxylic acid, 1-Ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
Beilstein:
750515
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

USP

mp

227-229 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN1C=C(C(O)=O)C(=O)c2ccc(C)nc12

InChI

1S/C12H12N2O3/c1-3-14-6-9(12(16)17)10(15)8-5-4-7(2)13-11(8)14/h4-6H,3H2,1-2H3,(H,16,17)

InChI key

MHWLWQUZZRMNGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George X Song-Zhao et al.
Mucosal immunology, 7(4), 763-774 (2013-11-28)
Polymorphisms in the intracellular pattern recognition receptor gene NLRP3 (NLR family, pyrin domain containing 3) have been associated with susceptibility to Crohn's disease, a type of inflammatory bowel disease. Following tissue damage or infection, NLRP3 triggers the formation of inflammasomes
Christopher M Parry
Transactions of the Royal Society of Tropical Medicine and Hygiene, 98(7), 413-422 (2004-05-13)
Multidrug-resistant (MDR) Salmonella Typhi (resistant to chloramphenicol, ampicillin, and trimethoprim-sulphamethoxazole) and isolates with reduced susceptibility to fluoroquinolones (indicated by resistance to nalidixic acid, NaR) have caused epidemics and become endemic in southern Viet Nam during the 1990s. Short courses of
Bing Gu et al.
International journal of antimicrobial agents, 40(1), 9-17 (2012-04-10)
Shigella is becoming an increasing public health problem due to development of multiple antimicrobial resistance, frequently resulting in treatment failure. A systematic review was conducted based on a literature search of computerised databases. Random or fixed-effects models were used, based
Solomon A Yimer et al.
Journal of clinical microbiology, 53(4), 1301-1309 (2015-02-13)
Recent genotyping studies of Mycobacterium tuberculosis in Ethiopia have reported the identification of a new phylogenetically distinct M. tuberculosis lineage, lineage 7. We therefore investigated the genetic diversity and association of specific M. tuberculosis lineages with sociodemographic and clinical parameters
Drug therapy reviews: nalidixic acid.
R Gleckman et al.
American journal of hospital pharmacy, 36(8), 1071-1076 (1979-08-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service