Skip to Content
Merck
All Photos(1)

Key Documents

1443001

USP

Metyrapone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Methyl-1,2-di-3-pyridyl-1-propanone, Metyrapone, Su-4885

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$570.00

$570.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
200 MG
$570.00

About This Item

Empirical Formula (Hill Notation):
C14H14N2O
CAS Number:
Molecular Weight:
226.27
Beilstein:
163023
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$570.00


Estimated to ship on26 May 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

metyrapone

manufacturer/tradename

USP

mp

52-55 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)(c1cccnc1)C(=O)c2cccnc2

InChI

1S/C14H14N2O/c1-14(2,12-6-4-8-16-10-12)13(17)11-5-3-7-15-9-11/h3-10H,1-2H3

InChI key

FJLBFSROUSIWMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metyrapone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam C Kulp et al.
Behavioural brain research, 382, 112480-112480 (2020-01-19)
Following a stressful event, the hypothalamus-pituitary-adrenal axis mediates the release of the stress hormone cortisol (corticosterone in rodents; CORT). Elevated CORT binds to glucocorticoid receptors to mediate physiological responses including facilitating memory formation. Previous work from our laboratory demonstrated that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service