Skip to Content
Merck
All Photos(1)

Key Documents

1374340

USP

Magnesium stearate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Stearic acid magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16CO2]2Mg
CAS Number:
Molecular Weight:
591.24
Beilstein:
3919702
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

manufacturer/tradename

USP

mp

200 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Mg]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Mg/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

HQKMJHAJHXVSDF-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service