Skip to Content
Merck
All Photos(1)

Key Documents

1368008

USP

Liothyronine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,3′,5-Triiodo-L-thyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine, Liothyronine, T3

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$606.00

$606.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$606.00

About This Item

Empirical Formula (Hill Notation):
C15H12I3NO4
CAS Number:
Molecular Weight:
650.97
Beilstein:
2710227
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$606.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

liothyronine

manufacturer/tradename

USP

mp

234-238 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C(O)=O

InChI

1S/C15H12I3NO4/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22/h1-4,6,12,20H,5,19H2,(H,21,22)/t12-/m0/s1

InChI key

AUYYCJSJGJYCDS-LBPRGKRZSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Liothyronine USP reference standard for use in specified quality tests and assays.

Also used to prepare standard, standard stock, Liothyronine stock, and resolution solutions for assay, and identification test according to given below monographs of the United States Pharmacopeia (USP):
  • Thyroid[1]
  • Levothyroxine Sodium Tablets[2]
  • Liothyronine Sodium Tablets[3]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Thyroid,Cardio-vascular system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liothyronine Sodium Tablets
United States Pharmacopeia, 34(4), 2640-2640 (2013)
Levothyroxine Sodium Tabletsl
United States Pharmacopeia, 42(2), 2618-2618 (2022)
Thyroid
United States Pharmacopeia, 40(5), 4362-4362 (2023)
Akihiro Mouri et al.
Psychoneuroendocrinology, 48, 147-161 (2014-07-13)
Attention deficit/hyperactivity disorder (ADHD) has been reported in association with resistance to thyroid hormone, a disease caused by a mutation in the thyroid hormone receptor β (TRβ) gene. TRβ is a key protein mediating down-regulation of thyrotropin (TSH) expression by
Sara Santos Bernardes et al.
Cell and tissue research, 357(3), 803-814 (2014-05-21)
The role of reactive oxygen species (ROS) in muscle protein hydrolysis and protein oxidation in thyrotoxicosis has not been explored. This study indicates that ROS play a role in skeletal muscle wasting pathways in thyrotoxicosis. Two experimental groups (rats) were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service