Skip to Content
Merck
All Photos(1)

Key Documents

1356552

USP

Ketoprofen Related Compound C

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-[(1RS)-1-Carboxyethyl]benzoic acid, 2-(3-Carboxyphenyl)propionic acid, 3-Carboxy-α-methylbenzeneacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$2,140.00

$2,140.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 MG
$2,140.00

About This Item

Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$2,140.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

ketoprofen

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)C(C)c1cc(ccc1)C(=O)O

InChI

1S/C10H10O4/c1-6(9(11)12)7-3-2-4-8(5-7)10(13)14/h2-6H,1H3,(H,11,12)(H,13,14)

InChI key

RIXOZEBXQQUHQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ketoprofen Related Compound C USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Ketoprofen Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service