Skip to Content
Merck
All Photos(1)

Key Documents

1354954

USP

Kaempferol-3-O-Robinoside-7-O-Glucoside

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Kaempferol 3-O-(6′′-O-α-L-Rhamnopyranosyl)-β-D-galactopyranoside7-O-β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$497.00

$497.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
20 MG
$497.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$497.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

pharmaceutical primary standard

Agency

certified by the USP

mol wt

756.66 (C33H40O20)

packaging

pkg of 20 mg

manufacturer/tradename

USP 1354954

application(s)

pharmaceutical

format

neat

storage temp.

room temp

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Kaempferol-3-O-Robinoside-7-O-Glucoside USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Ashwagandha Root Powder
  • Ashwagandha Root Dry Extract

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

USP issued SDS can be found here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service