Skip to Content
Merck
All Photos(1)

Key Documents

1161000

USP

Cyproheptadine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Cyproheptadine hydrochloride sesquihydrate, 4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methyl-piperidine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$606.00

$606.00


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$606.00

About This Item

Empirical Formula (Hill Notation):
C21H21N · HCl · 1.5H2O
CAS Number:
Molecular Weight:
350.88
UNSPSC Code:
41116107
NACRES:
NA.24

$606.00


Estimated to ship on29 March 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

cyproheptadine

manufacturer/tradename

USP

mp

165 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O.O.Cl.CN1CCC(\CC1)=C2\c3ccccc3C=Cc4ccccc24.CN5CCC(\CC5)=C6\c7ccccc7C=Cc8ccccc68

InChI

1S/2C21H21N.ClH.3H2O/c2*1-22-14-12-18(13-15-22)21-19-8-4-2-6-16(19)10-11-17-7-3-5-9-20(17)21;;;;/h2*2-11H,12-15H2,1H3;1H;3*1H2

InChI key

PXCSXCDWTUPVIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cyproheptadine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Cyproheptadine Hydrochloride Oral Solution
  • Cyproheptadine Hydrochloride Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service