Skip to Content
Merck
All Photos(1)

Key Documents

1154015

USP

Cyclizine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1-Methylpiperazine

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$1,390.00

$1,390.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$1,390.00

About This Item

Empirical Formula (Hill Notation):
C5H12N2
CAS Number:
Molecular Weight:
100.16
Beilstein:
102724
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,390.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

vapor density

3.5 (vs air)

API family

cyclizine

manufacturer/tradename

USP

refractive index

n20/D 1.466 (lit.)

bp

138 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1CCNCC1

InChI

1S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3

InChI key

PVOAHINGSUIXLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cyclizine Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Cyclizine Hydrochloride
  • Cyclizine Hydrochloride Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

88.7 °F - closed cup

Flash Point(C)

31.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service