Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
15 mg | Check Cart for Availability | $2,080.00 |
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
grade
pharmaceutical primary standard
agency
USP
API family
betamethasone
manufacturer/tradename
USP
application(s)
pharmaceutical
format
mixture
storage temp.
2-8°C
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
USP issued SDS can be found here.
USP issued SDS can be found here.
Application
Betamethasone Valerate System Suitability Mixture USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
A mixture containing betamethasone valerate and betamethasone valerate related compound H
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade - |
| format mixture | format neat | format mixture | format neat |
| application(s) pharmaceutical | application(s) pharmaceutical | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecules) |
| API family betamethasone | API family everolimus | API family carvedilol | API family nilotinib |
| agency USP | agency USP | agency - | agency - |
signalword
Danger
hcodes
Hazard Classifications
Repr. 1B - STOT RE 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


