Skip to Content
Merck
All Photos(1)

Key Documents

1000829

USP

Abiraterone System Suitability Mixture

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
$2,660.00

$2,660.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
20 MG
$2,660.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,660.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

pharmaceutical primary standard

API family

abiraterone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

mixture

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Abiraterone System Suitability Mixture USP reference standard suitable for use in specified USP compendial tests and assays.

Also used to prepare system suitability solution during the assay of Abiraterone Acetate and Abiraterone Acetate Tablets by using liquid chromatography coupled to a UV detector. [1] [2]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Repr. 1B - STOT RE 2

Target Organs

Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abiraterone Acetate
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(6), 25-25 (2020)
Abiraterone Acetate Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(6), 26-26 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service