Skip to Content
Merck
All Photos(1)

Key Documents

TA9H97BAF25C

Targetmol Chemicals Inc.

Pinealon Acetate

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
478.46

Iupac Name

(S)-4-amino-5-(((S)-3-carboxy-1-(((S)-1-carboxy-4-((diaminomethylene)amino)butyl)amino)-1-oxopropan-2-yl)amino)-5-oxopentanoic acid--acetic acid (1/1)

Inchi Code

1S/C15H26N6O8.C2H4O2/c16-7(3-4-10(22)23)12(26)21-9(6-11(24)25)13(27)20-8(14(28)29)2-1-5-19-15(17)18;1-2(3)4/h7-9H,1-6,16H2,(H,20,27)(H,21,26)(H,22,23)(H,24,25)(H,28,29)(H4,17,18,19);1H3,(H,3,4)/t7-,8-,9-;/m0./s1

InChI key

GDKWZMZURALPEA-YWUTZLAHSA-N

storage temp.

-80/-20

Purity

99.55%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Pinealon Acetate

MSDS

https://www.targetmol.com/compound/Pinealon Acetate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library