Skip to Content
Merck
All Photos(1)

Documents

Z777292

Antibodix® WCX-NP (5 μm) Weak Cation Exchange HPLC Columns

L × I.D. 5 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Antibodix® WCX-NP5 Weak Cation Exchange Column, L × I.D. 5 cm × 4.6 mm, 5 μm particle size

material

stainless steel

product line

Antibodix®

manufacturer/tradename

Sepax 602NP5-4605

packaging

1 ea of

parameter

40 Mpa max. pressure (6000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

5 cm × 4.6 mm

matrix

polymeric

matrix active group

weak cation exchange phase

particle size

5 μm

pore size

non-porous

operating pH range

2.0-12.0

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

The non-porous poly(styrene divinylbenzene) particles in Antibodix columns are shielded by a hydrophilic polymer layer that contains a network of weak (WCX) cation exchange groups. Antibodix columns are designed for the reproducible analysis of charge variants of monoclonal antibodies by salt or pH gradient.

Antibodix User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Antibodix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service