Skip to Content
Merck
All Photos(1)

Key Documents

Z777114

SRT®-C SEC-500 (5 µm) HPLC Columns

L × I.D. 30 cm × 4.6 mm, stainless steel

Synonym(s):

TM=["SRT"] -C SEC-500 Size Exclusion Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$2,550.00

$2,550.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$2,550.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$2,550.00


Check Cart for Availability

Product Name

SRT®-C SEC-500 Gel Filtration Column, 5 μm, L × I.D. 30 cm × 4.6 mm, 5 μm particle size, stainless steel

material

stainless steel

Agency

suitable for USP L59

product line

SRT®

mol wt

15,000-5,000,000

manufacturer/tradename

Sepax 235500-4630

packaging

1 ea of

parameter

20 Mpa max. pressure (3000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

30 cm × 4.6 mm

impurities

<10 ppm metals

matrix

spherical silica
fully porous particle

matrix active group

hydrophilic polymer phase

particle size

5 μm

pore size

500 Å

operating pH range

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

The bonded phase in SRT-C SEC columns is more polar than the hydrophilic phase used in SRT columns, making SRT-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies. SRT-C columns share with SRT columns the same 5 micron silica particle and the same wide range of pore sizes so you can select the pore size that will allow full access for your compound of interest.

SRT-C User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SRT is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service