Z777092
Zenix®-C SEC-300 (3 µm) HPLC Columns
L x I.D. 30 cm × 7.8 mm, hydrophilic polymer phase
About This Item
Recommended Products
product name
Zenix®-C SEC-300 Gel Filtration Column, 3 μm, L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
material
stainless steel
Agency
suitable for USP L59
product line
Zenix®
mol wt
5,000-1,250,000
manufacturer/tradename
Sepax 233300-7830
packaging
1 ea of
parameter
20 Mpa max. pressure (3000 psi)
80 °C max. temp.
technique(s)
HPLC: suitable
L × I.D.
30 cm × 7.8 mm
impurities
<10 ppm metals
matrix
spherical silica
fully porous particle
matrix active group
hydrophilic polymer phase
particle size
3 μm
pore size
300 Å
operating pH range
2.0-8.5
separation technique
size exclusion (SEC)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Zenix User Manual
Application
- Quantitative analysis of nine types of virus-like particles in human papilloma virus bulk by size-exclusion chromatography: This study conducted a quantitative analysis of nine types of virus-like particles (VLPs) in human papilloma virus (HPV) bulk using size-exclusion chromatography (SEC). The Zenix-C SEC-300 Gel Filtration Column, 3 m mu m, was utilized to achieve high-resolution separation of the VLPs. The application of this gel filtration column enabled accurate and reproducible quantification of the VLPs, which is crucial for the quality control of HPV vaccines. The research highlights the effectiveness of the Zenix-C SEC-300 column in biopharmaceutical analysis and quality assurance (Long et al., 2021).
Legal Information
Application
antibody
guard column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Characterize mAb monomers, aggregates, and fragments using SEC-UV workflow with Zenix® and Zenix®-C SEC columns.
Chromatograms
application for HPLCapplication for HPLCapplication for HPLCapplication for HPLCShow MoreOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service