Skip to Content
Merck
All Photos(1)

Key Documents

Z777065

SRT® SEC-1000 (5 µm) HPLC Columns

L × I.D. 30 cm × 4.6 mm, PEEK

Synonym(s):

TM=["SRT"] SEC-1000 Size Exclusion Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$2,690.00

$2,690.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$2,690.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$2,690.00


Check Cart for Availability

Product Name

SRT® SEC-1000 Gel Filtration Column, 5 μm, L × I.D. 30 cm × 4.6 mm, 5 μm particle size, PEEK

material

PEEK

Agency

suitable for USP L59

product line

SRT®

mol wt

50,000-7,500,000

manufacturer/tradename

Sepax 215950P-4630

packaging

1 ea of

parameter

20 Mpa max. pressure (3000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

30 cm × 4.6 mm

impurities

<10 ppm metals

matrix

spherical silica
fully porous particle

matrix active group

hydrophilic polymer phase

particle size

5 μm

pore size

1000 Å

operating pH range

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Sepax SRT SEC columns feature a hydrophilic surface layer that effectively shields the highly porous, five micron silica particles from interacting with your sample. SRT columns are employed in the quality control of biotherapeutics by gel filtration chromatography. Select from a wide variety of available pore sizes to analyze peptides, proteins, glycoproteins, vaccines and water soluble polymers.

SRT User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SRT is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Z777065, SRT® SEC-1000 (5 µm) HPLC Columns. Can we use water as the mobile phase for the analysis of molecular weight?

    1 answer
    1. The SRT SEC columns are compatible with an aqueous mobile phase or a mixture of organic solvent and water, such as methanol or acetonitrile and water. Therefore, water can be used for the analysis of the molecular weight of polar polymers.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service