Skip to Content
Merck
All Photos(1)

Documents

Y0002381

Supelco

Propylthiouracil impurity A

CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Thiourea, Sulfourea, Thiocarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSNH2
CAS Number:
Molecular Weight:
76.12
Beilstein:
605327
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

propylthiouracil

manufacturer/tradename

EDQM

mp

170-176 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

NC(N)=S

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)

InChI key

UMGDCJDMYOKAJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. Compendial materials should be used for the purpose described in the appropriate monograph unless otherwise authorized by the National Competent Authority. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins

Packaging

Unit quantity: 15mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service