Skip to Content
Merck
All Photos(1)

Documents

Y0002292

Ciclosporin for system suitability A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cyclosporin A, Antibiotic S 7481F1, Cyclosporine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H111N11O12
CAS Number:
Molecular Weight:
1202.61
Beilstein:
3647785
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

API family

ciclosporin

manufacturer/tradename

EDQM

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C

InChI

1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1

InChI key

PMATZTZNYRCHOR-CGLBZJNRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.

Application

Ciclosporin for system suitability A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service