SU860012
Vials, screw top, clear glass, 1.5 mL
volume 1.5 mL, clear glass vial, thread for 10-425, O.D. × H × I.D. 11.6 mm × 32 mm × 6 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
Recommended Products
material
clear glass vial
feature
closure type screw top vial
packaging
pkg of 1000 ea
O.D. × H × I.D.
11.6 mm × 32 mm × 6 mm
volume
1.5 mL
fitting
thread for 10-425
Looking for similar products? Visit Product Comparison Guide
Other Notes
27265 --> material nos. 27265 (100pk) & 27266 (2000pk).
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service