Skip to Content
Merck
All Photos(1)

Key Documents

PS92-250MG

Ethion

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H22O4P2S4
CAS Number:
Molecular Weight:
384.48
Beilstein:
1804530
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-92

SMILES string

CCOP(=S)(OCC)SCSP(=S)(OCC)OCC

InChI

1S/C9H22O4P2S4/c1-5-10-14(16,11-6-2)18-9-19-15(17,12-7-3)13-8-4/h5-9H2,1-4H3

InChI key

RIZMRRKBZQXFOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L John R Foster et al.
FEMS microbiology letters, 240(1), 49-53 (2004-10-27)
The organophosphate pesticide, Ethion, remains a major environmental contaminant in rural Australia and poses a significant threat to environmental and public health. The aerobic degradation of Ethion by mesophilic bacteria isolated from contaminated soils surrounding disused cattle dip sites was
Huilong Xia et al.
Bioresource technology, 97(8), 1050-1054 (2005-06-29)
The potential of water hyacinth (Eichhornia crassipes) to remove a phosphorus pesticide ethion were investigated. The disappearance rate constants of ethion in culture solutions were 0.01059, 0.00930, 0.00294, and 0.00201 h-1 for the non-sterile planted, sterile planted, non-sterile unplanted, and
Riaz Ahmad et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 39(4), 565-576 (2004-10-12)
The potential of five nonionic surfactants, Triton X-100, Brij35, Ethylan GE08, Ethylan CD127, and Ethylan CPG660 for enhancing release of carbaryl and ethion from two long-term contaminated soils was evaluated using the batch method. Incorporation of the surfactants into soils
H B Christensen et al.
Food additives and contaminants, 20(8), 764-775 (2003-09-18)
The estimation of uncertainty of an analytical result has become important in analytical chemistry. It is especially difficult to determine uncertainties for multiresidue methods, e.g. for pesticides in fruit and vegetables, as the varieties of pesticide/commodity combinations are many. In
G K Bhatti et al.
Cellular and molecular biology (Noisy-le-Grand, France), 57(1), 70-79 (2011-03-04)
Erythrocyte membranes are an excellent model system to study interaction of pro-oxidants with membranes. The aim of the present study is to examine the effect of vitamin E on ethion-induced biochemical and morphological alterations in erythrocytes. Ethion was administered to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service