Skip to Content
Merck
All Photos(1)

Documents

PS90-250MG

Diazinon

analytical standard

Synonym(s):

Dimpylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21N2O3PS
CAS Number:
Molecular Weight:
304.35
Beilstein:
273790
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-90

SMILES string

CCOP(=S)(OCC)Oc1cc(C)nc(n1)C(C)C

InChI

1S/C12H21N2O3PS/c1-6-15-18(19,16-7-2)17-11-8-10(5)13-12(14-11)9(3)4/h8-9H,6-7H2,1-5H3

InChI key

FHIVAFMUCKRCQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Wang et al.
Reproductive toxicology (Elmsford, N.Y.), 34(4), 489-497 (2012-09-05)
The potential toxicity resulting from combinatorial effects of organophosphorus and pyrethroid insecticides are not completely known. We evaluated male reproductive toxicity in mice co-exposed to diazinon and cis-permethrin. Nine-week-old male Sv/129 mice were exposed to diazinon (10 μmol/kg/day) or cis-permethrin
M J van der Schans et al.
Chemico-biological interactions, 203(1), 96-102 (2012-11-06)
Organophosphate (OP) pesticides are neurotoxic compounds that are widely used in agriculture. Classical methods for monitoring OP exposure comprise the measurement of intact OP, its metabolites or cholinesterase activity. Newly developed methods focus on the analysis of the OP adduct
K P Shyma et al.
Experimental & applied acarology, 58(4), 471-481 (2012-07-05)
The multi host tick, Hyalomma anatolicum, is the commonest Hyalomma species in India and cattle serves as the main host of this species. A study to evaluate the acaricide resistance of H. anatolicum to deltamethrin, cypermethrin and diazinon was conducted
Xiao Zhang et al.
Environmental toxicology and pharmacology, 34(3), 959-968 (2012-09-12)
Pesticide exposure has repeatedly been associated with cancers. However, molecular mechanisms are largely undetermined. In this study, we examined whether exposure to diazinon, a common organophosphate that has been associated with cancers, could induce DNA methylation alterations. We conducted genome-wide
Karen Huen et al.
Environmental research, 117, 8-16 (2012-06-12)
Organophosphate pesticides are widely used and recent studies suggest associations of in utero exposures with adverse birth outcomes and neurodevelopment. Few studies have characterized organophosphate pesticides in human plasma or established how these levels correlate to urinary measurements. We measured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service