Skip to Content
Merck
All Photos(1)

Documents

PS665

Abate®

analytical standard

Synonym(s):

Temephos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20O6P2S3
CAS Number:
Molecular Weight:
466.47
Beilstein:
1896901
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-665

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COP(=S)(OC)Oc1ccc(Sc2ccc(OP(=S)(OC)OC)cc2)cc1

InChI

1S/C16H20O6P2S3/c1-17-23(25,18-2)21-13-5-9-15(10-6-13)27-16-11-7-14(8-12-16)22-24(26,19-3)20-4/h5-12H,1-4H3

InChI key

WWJZWCUNLNYYAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Abate is a registered trademark of BASF

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clare Strode et al.
PloS one, 7(8), e39439-e39439 (2012-08-08)
The mosquito Aedes aegypti is one of the most important disease vectors because it transmits two major arboviruses, dengue and yellow fever, which cause significant global morbidity and mortality. Chemical insecticides form the cornerstone of vector control. The organophosphate temephos
Andrea Gómez et al.
Memorias do Instituto Oswaldo Cruz, 106(8), 993-996 (2012-01-14)
A susceptible strain of Aedes albopictus derived from the Gainesville strain (Florida, USA) was established in our laboratory. The larvicidal efficacies of the neurotoxic insecticides temephos, permethrin and the pure cis and trans-permethrin isomers and the microbial insecticide Bacillus thuringiensis
Seong Chun Chae et al.
Journal of medical entomology, 51(4), 817-823 (2014-08-15)
Culex pipiens molestus Forskal (Diptera: Culicidae) is the dominant mosquito species in septic tanks in South Korea. An assessment was made of the biological control potential of mud loaches, Misgurnus mizolepis Günther (Cypriniformes: Cobitidae), toward Cx. p. molestus larvae in
Argentina A Garza-Robledo et al.
Journal of the American Mosquito Control Association, 27(4), 404-407 (2012-02-15)
The effectiveness of spinosad and temephos for the control of mosquito larvae was evaluated in a tire dump in Allende, Nuevo Leon, Mexico. Three groups of 12 to 17 tires located in tree shade were utilized for this study. After
Mirna Velki et al.
Chemosphere, 90(11), 2690-2696 (2013-01-10)
Despite its negative effects, formalin has been often used for the expulsion of earthworms due to its high efficiency; however it is not known whether it will affect any significant measurable molecular processes in sampled earthworms. The aim of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service