PHR9188
Metoprolol EP Impurity G
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-(4-Hydroxyphenyl)ethanol, 4-Hydroxyphenethyl alcohol, Tyrosol, p-HPEA
Select a Size
$841.00
Select a Size
About This Item
$841.00
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
Agency
Not traceable to any primary pharmacopoeia standard
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
mp
89-92 °C (lit.)
application(s)
pharmaceutical small molecule
storage temp.
−20°C
SMILES string
OCCc1ccc(O)cc1
InChI
1S/C8H10O2/c9-6-5-7-1-3-8(10)4-2-7/h1-4,9-10H,5-6H2
InChI key
YCCILVSKPBXVIP-UHFFFAOYSA-N
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Biochem/physiol Actions
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service