Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥98.0% (HPLC)
form
crystals
manufacturer/tradename
PhytoLab
mp
222 °C (dec.) (lit.)
SMILES string
[H][C@]12CC=C3C4CC(C)(C)CC[C@@]4(CC[C@@]3(C)[C@]1(C)CCC5[C@](C)(CO)[C@H](CC[C@]25C)O[C@@H]6OC[C@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@@H](C)[C@H](O)[C@@H](O)[C@H]7O)C(=O)O[C@@H]8O[C@H](CO[C@@H]9O[C@H](CO)[C@@H](O[C@@H]%10O[C@@H](C)[C@H](O)[C@@H](O)[C@H]%10O)[C@H](O)[C@H]9O)[C@@H](O)[C@H](O)[C@H]8O
InChI
1S/C59H96O26/c1-24-34(63)38(67)42(71)49(78-24)83-46-29(20-60)80-48(45(74)41(46)70)77-22-30-37(66)40(69)44(73)51(81-30)85-53(75)59-17-15-54(3,4)19-27(59)26-9-10-32-55(5)13-12-33(56(6,23-61)31(55)11-14-58(32,8)57(26,7)16-18-59)82-52-47(36(65)28(62)21-76-52)84-50-43(72)39(68)35(64)25(2)79-50/h9,24-25,27-52,60-74H,10-23H2,1-8H3/t24-,25-,27?,28-,29+,30+,31?,32+,33-,34-,35-,36-,37+,38+,39+,40-,41+,42+,43+,44+,45+,46+,47+,48+,49-,50-,51-,52-,55-,56-,57+,58+,59-/m0/s1
InChI key
RYHDIBJJJRNDSX-WBOYGZCJSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Cholinergic Pathway Interaction in Gastrointestinal Research: Hederacoside C has been studied for its role in inducing contractions in isolated rat stomach strips through cholinergic pathways, providing insights into its potential pharmacological effects on gastrointestinal motility (Mendel et al., 2012).
- Pharmacological Effects on Gastrointestinal Activity: The compound′s effects have been further explored in the context of the whole extract of Hedera helix leaves, suggesting potential uses in treating conditions related to gastrointestinal dysmotility (Mendel et al., 2011).
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service