Skip to Content
Merck
All Photos(1)

Key Documents

PHL86249

Tetrahydropiperine

phyproof® Reference Substance

Synonym(s):

5-(1,3-Benzodioxol-5-yl)-1-(1-piperidinyl)-1-pentanone

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$621.00

$621.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$621.00

About This Item

Empirical Formula (Hill Notation):
C17H23NO3
CAS Number:
Molecular Weight:
289.37
MDL number:
UNSPSC Code:
12352210
NACRES:
NA.24

$621.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0%

manufacturer/tradename

PhytoLab

application(s)

food and beverages
pharmaceutical

SMILES string

N3(CCCCC3)C(=O)CCCCc1cc2c(cc1)OCO2

InChI key

APZYKUZPJCQGPP-UHFFFAOYSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service