Skip to Content
Merck
All Photos(1)

Key Documents

PHL85776

α-Spinasterol

phyproof® Reference Substance

Synonym(s):

α-Spinasterin, (3β,5α,22E)-Stigmasta-7,22-dien-3-ol, (E)-5α-Stigmasta-7,22-dien-3β-ol, 5α-Stigmasta-7,22-dien-3β-ol, Bessisterol, Hitodesterol, Spinasterol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$827.00

$827.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$827.00

About This Item

Empirical Formula (Hill Notation):
C29H48O
CAS Number:
Molecular Weight:
412.69
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$827.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

C[C@@]12[C@](CC[C@]2([H])[C@H](C)/C=C/[C@@H](CC)C(C)C)([H])C3=CC[C@@]4([H])C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC1

InChI

1S/C29H48O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h8-9,11,19-23,25-27,30H,7,10,12-18H2,1-6H3/b9-8+/t20-,21-,22+,23+,25-,26+,27+,28+,29-/m1/s1

InChI key

JZVFJDZBLUFKCA-FXIAWGAOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

α-Spinasterol is a blood-brain barrier-permeable and orally active plant phytosterol with antioxidant, anticonvulsant (0.1-1 mg/kg i.p. in mice), antidepressant (1-2 mg/kg i.p. in mice) and anti-inflammatory (0.001-1 mg/kg, i.p. or 1-10 mg/kg, i.g. in mice) properties, as well as antinociceptive efficacy via dual actions against transient receptor potential vanilloid 1 (TrpV1; IC50/Emax = 1.4 μM/67% against 2 nM RTX for binding mouse spinal cord membranes; IC50/Emax = 40 μM/62% against 20 μM capsaicin-induced mouse spinal cord synaptosomes Ca2+ influx) and cyclooxygenases (IC50 = 16.17 μM/COX1 and 7.76μM/COX2). a-spinasterol can directly affect membrane structure and packing in a manner similar to cholesterol, both a-spinasterol and stitosterol are reported to lower plasma/liver cholesterol levels and increase fecal cholesterol excretion when supplemented in daily diet (1%) in mice in vivo.
Orally active phytosterol with anticonvulsant, antidepressant, anti-inflammatory, and antinociceptive efficacy via dual actions against TrpV1 and COX1/2.

Other Notes

This compound is commonly found in plants of the genus: hedera

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service