Skip to Content
Merck
All Photos(1)

Key Documents

PHL84103

Riddelliine N-oxide

phyproof® Reference Substance

Synonym(s):

Riddeline N-oxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$782.00

$782.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$782.00

About This Item

Empirical Formula (Hill Notation):
C18H23NO7
CAS Number:
Molecular Weight:
365.38
UNSPSC Code:
41116107
NACRES:
NA.24

$782.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

−20°C

SMILES string

[N+]21([C@H]3[C@H](OC(=O)\C(=C/C)\CC(=C)[C@](C(=O)OCC3=CC2)(O)CO)CC1)[O-]

InChI

1S/C18H23NO7/c1-3-12-8-11(2)18(23,10-20)17(22)25-9-13-4-6-19(24)7-5-14(15(13)19)26-16(12)21/h3-4,14-15,20,23H,2,5-10H2,1H3/b12-3-/t14-,15-,18-,19?/m1/s1

InChI key

NPENPMDVCQGSSO-AHPXGCJSSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Analysis Note

For the analysis of the phytotoxins. Pyrrolizidin alkaloid (PA)

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2 - Repr. 2 - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service