Skip to Content
Merck
All Photos(1)

Documents

PHL82581

(+)-Longifolene

phyproof® Reference Substance

Synonym(s):

(+)-Longifolene, (1R,2S,7S,9S)-3,3,7-Trimethyl-8-methylenetricyclo[5.4.0.02.9]undecane, Junipene, Kuromatsuene, [1S-(1α,3aβ,4α,8aβ)]-Decahydro-4,8,8-trimethyl-9-methylene-1,4-methanoazulene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
2044263
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥75.0% (GC)

manufacturer/tradename

PhytoLab

refractive index

n20/D 1.504 (lit.)

bp

254 °C/706 mmHg (lit.)

density

0.928 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)CCC[C@]2(C)C3CCC(C13)C2=C

InChI

1S/C15H24/c1-10-11-6-7-12-13(11)14(2,3)8-5-9-15(10,12)4/h11-13H,1,5-9H2,2-4H3/t11?,12?,13?,15-/m0/s1

InChI key

PDSNLYSELAIEBU-WOFVOEOOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service