Skip to Content
Merck
All Photos(1)

Key Documents

PHL80966

(+)-Dihydrocinchonine

phyproof® Reference Substance

Synonym(s):

Hydrocinchonine, Dihydrocinchonine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$641.00

$641.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$641.00

About This Item

Empirical Formula (Hill Notation):
C19H24N2O
CAS Number:
Molecular Weight:
296.41
Beilstein:
4296749
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$641.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

mp

269-272 °C (lit.)

SMILES string

CC[C@H]1CN2CC[C@H]1C[C@@H]2[C@@H](O)c3ccnc4ccccc34

InChI

1S/C19H24N2O/c1-2-13-12-21-10-8-14(13)11-18(21)19(22)16-7-9-20-17-6-4-3-5-15(16)17/h3-7,9,13-14,18-19,22H,2,8,10-12H2,1H3/t13-,14-,18+,19-/m0/s1

InChI key

WFJNHVWTKZUUTR-QAMTZSDWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service