Skip to Content
Merck
All Photos(1)

Key Documents

PBG7A

Supelco

LEAD IN GASOLINE

analytical standard, set of 7 × 100 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

Agency

ASTM® D5059 (Part A)

packaging

set of 7 × 100 mL

technique(s)

AAS: suitable
ICP: suitable

application(s)

petroleum

format

matrix material

storage temp.

room temp

General description

7 standards per set; 100 mL each.

Analysis Note

Lead Standards in Various Matrices - Formulated sets
Standard No.Lead (Gms/Gal.)

Concentration Table: PBG7A
1 . . . . . . . . .0.000
2 . . . . . . . . .0.100
3 . . . . . . . . .1.000
4 . . . . . . . . .2.000
5 . . . . . . . . .3.000
6 . . . . . . . . .4.000
7 . . . . . . . . .5.000

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 1 - Lact. - Muta. 1B - Repr. 1A - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system,Blood,Immune system,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.0 °F - closed cup

Flash Point(C)

18.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service