Skip to Content
Merck
All Photos(1)

Key Documents

N12729

Oryzalin

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N4O6S
CAS Number:
Molecular Weight:
346.36
Beilstein:
2177305
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1 g

manufacturer/tradename

Chem Service, Inc. N-12729-1G

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CCCN(CCC)c1c(cc(cc1[N+]([O-])=O)S(N)(=O)=O)[N+]([O-])=O

InChI

1S/C12H18N4O6S/c1-3-5-14(6-4-2)12-10(15(17)18)7-9(23(13,21)22)8-11(12)16(19)20/h7-8H,3-6H2,1-2H3,(H2,13,21,22)

InChI key

UNAHYJYOSSSJHH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Blood,Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuuta Imoto et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(23), 9583-9588 (2013-05-23)
Peroxisomes (microbodies) are ubiquitous single-membrane-bounded organelles and fulfill essential roles in the cellular metabolism. They are found in virtually all eukaryotic cells and basically multiply by division. However, the mechanochemical machinery involved in peroxisome division remains elusive. Here, we first
Mathias Sorieul et al.
Traffic (Copenhagen, Denmark), 12(11), 1552-1562 (2011-08-02)
We screened a panel of compounds derived from Exo2 - a drug that perturbs post-Golgi compartments and trafficking in mammalian cells - for their effect on the secretory pathway in Arabidopsis root epidermal cells. While Exo2 and most related compounds
Atsuko Era et al.
Journal of plant research, 126(1), 113-119 (2012-06-09)
Actin microfilaments play crucial roles in diverse plant functions. Some specific cellular processes require interaction between F-actin and microtubules, and it is believed that there are direct or indirect connections between F-actin and microtubules. We previously reported that actin microfilaments
William Eisinger et al.
Molecular plant, 5(3), 601-610 (2012-03-10)
Radially arranged cortical microtubules are a prominent feature of guard cells. Guard cells expressing GFP-tubulin showed consistent changes in the appearance of microtubules when stomata opened or closed. Guard cells showed fewer microtubule structures as stomata closed, whether induced by
Stefano Morettini et al.
Plant physiology and biochemistry : PPB, 63, 107-114 (2012-12-25)
Sensitivity to the two anti-microtubular drugs oryzalin and EPC (ethyl-N-phenylcarbamate) is shown to be uncoupled in the rice EPC-resistant ER31d cell line, derived from the corresponding ER31 mutant. The ER31d cell line grows in the presence of EPC but it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service