Skip to Content
Merck
All Photos(1)

Key Documents

N12483

N-(2-Ethylhexyl)-5-norbornene-2,3-dicarboximide

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25NO2
CAS Number:
Molecular Weight:
275.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-962

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CCCCC(CC)CN1C(=O)C2[C@H]3C[C@H](C=C3)C2C1=O

InChI

1S/C17H25NO2/c1-3-5-6-11(4-2)10-18-16(19)14-12-7-8-13(9-12)15(14)17(18)20/h7-8,11-15H,3-6,9-10H2,1-2H3/t11?,12-,13+,14?,15?

InChI key

WLLGXSLBOPFWQV-OTHKPKEBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

350.6 °F - closed cup

Flash Point(C)

177 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H S Radwan et al.
Toxicology, 14(3), 193-198 (1979-11-01)
Piperonyl butoxide, sulfoxide and MGK 264 were found to act as synergists for certain organotin compounds; with respect to their toxicity on the larvae of a susceptible strain of the cotton leafworm, Spodoptera littoralis (Boisd.). High degree of synergism was
I H Wang et al.
Journal of chromatography. A, 864(2), 271-281 (2000-02-11)
N-Octyl bicycloheptene dicarboximide (MGK 264) has exo and endo diastereomers. Each structure has a chiral center at the nitrogen side chain. Enantioselective separation of MGK 264 was achieved by normal-phase high-performance liquid chromatography (HPLC) using cellulose-based Chiralcel OD column with
Effects of synergists on bendiocarb and pyrethrins resistance in the German cockroach (Dictyoptera: Blattellidae).
D G Cochran
Journal of economic entomology, 80(4), 728-732 (1987-08-01)
I-Hsiung Wang et al.
Journal of chromatography. A, 983(1-2), 145-152 (2003-02-06)
An isocratic reversed-phase high-performance liquid chromatographic (RP-HPLC) method development for the simultaneous determination of five active ingredients (S)-methoprene, MGK264, PBO, sumithrin and permethrin in a new complex pesticide formulation is described. These active ingredients have similar polarities and contain isomers.
G L Zellers et al.
Annals of emergency medicine, 19(3), 327-329 (1990-03-01)
A man experienced one hour and 40 minutes of continual, inappropriate, uncontrollable laughter. The onset was preceded by a single-inhalation exposure to an insecticide of very low toxicity. The episode was terminated by a single dose of IV diazepam. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service